Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083916.1 | NP_001077385.1 | 2 | 908 | ||
NM_133707.2 | NP_598468.2 | 2 | 849 | |||
GenBank mRNA | AK007551.1 | - | 1 | 774 | ||
AK158383.1 | - | 2 | 995 | |||
BC006890.1 | - | 2 | 777 | 3599314 |