Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026934.3 | NP_081210.2 | 10 | 1222 | ||
GenBank mRNA | AK007452.1 | - | 5 | 453 | ||
AK139511.1 | - | 10 | 1318 | |||
AK152374.1 | - | 10 | 1313 | |||
AK159596.1 | - | 10 | 1294 | |||
AK167696.1 | - | 10 | 1315 | |||
AK216553.1 | - | 2 | 137 | |||
BC028787.1 | - | 5 | 661 | 1246186 | ||
BC031845.1 | - | 10 | 1222 | 3983096 | ||
BC058229.1 | - | 10 | 1149 | 6313954 |