Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026935.4 | NP_081211.3 | 8 | 1049 | ||
GenBank mRNA | AK007384.1 | - | 8 | 909 | ||
AK144336.1 | - | 8 | 989 | |||
AY005469.1 | - | 8 | 1049 | |||
BC022665.1 | - | 8 | 1042 | 4223678 |