Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxa1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026936.3 | NP_081212.1 | 10 | 1199 | ||
GenBank mRNA | AK007567.1 | - | 10 | 1201 | ||
AK028379.1 | - | 10 | 1198 | |||
AK044817.1 | - | 10 | 1198 | |||
AK052432.1 | - | 10 | 1200 | |||
AK077624.1 | - | 10 | 1200 | |||
AK087756.1 | - | 10 | 1203 | |||
AK139484.1 | - | 10 | 1207 |