Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178027.4 | NP_821170.1 | 4 | 1096 | ||
GenBank mRNA | AK010160.1 | - | 4 | 1030 | ||
AK031589.1 | - | 4 | 1099 | |||
AK035321.1 | - | 4 | 1047 | |||
AK150929.1 | - | 4 | 1017 | |||
AK155578.1 | - | 4 | 1057 | |||
AK165539.1 | - | 4 | 1035 | |||
AK171154.1 | - | 4 | 1055 | |||
BC029758.1 | - | 2 | 189 | |||
BC049180.1 | - | 4 | 1059 | 6314347 | ||
BC052721.1 | - | 4 | 1020 | 6809853 | ||
BC062972.1 | - | 4 | 1020 | 6809878 |