Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ydjc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026940.4 | NP_081216.1 | 5 | 1013 | ||
GenBank mRNA | AK007503.1 | - | 5 | 1002 | ||
BC030425.1 | - | 3 | 1297 | |||
BC115584.1 | - | 5 | 1001 | 40102346 | ||
BC115585.1 | - | 5 | 1001 | 40102347 |