Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ydjc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026940.4 | NP_081216.1 | 2, 3 | 348 | ||
GenBank mRNA | AK007503.1 | - | 2, 3 | 337 | ||
AK032396.1 | - | 378 | ||||
AK140044.1 | - | 453 | ||||
AK141482.1 | - | 1 | 628 | |||
BC030425.1 | - | 2, 3 | 350 | |||
BC115584.1 | - | 2, 3 | 336 | 40102346 | ||
BC115585.1 | - | 2, 3 | 336 | 40102347 |