Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164336.1 | NP_001157808.1 | 5 | 992 | ||
NM_027314.3 | NP_081590.3 | 5 | 992 | |||
GenBank mRNA | AK003478.1 | - | 1 | 37 | ||
AK007511.1 | - | 5 | 828 | |||
AK009232.1 | - | 5 | 978 | |||
AK009364.1 | - | 5 | 990 | |||
AK205563.1 | - | 1 | 59 | |||
BC064752.1 | - | 5 | 716 | 30049608 | ||
BC107214.1 | - | 5 | 699 | |||
BC107215.1 | - | 5 | 704 | 40051628 | ||
BC132455.1 | - | 5 | 756 | 40130732 | ||
BC132457.1 | - | 5 | 755 | 40130734 |