Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026949.3 | NP_081225.1 | 6, 7 | 885 | ||
GenBank mRNA | AK007581.1 | - | 6, 7 | 949 | ||
AK075727.1 | - | 6, 7 | 855 | |||
AK150757.1 | - | 6, 7 | 966 | |||
AK153907.1 | - | 6, 7 | 888 | |||
AK192037.1 | - | 306 | ||||
AK192809.1 | - | 2, 3 | 222 | |||
AK199523.1 | - | 2, 3 | 212 | |||
AY515608.1 | - | 5, 6 | 851 | |||
BC004040.1 | - | 6, 7 | 885 | 3496188 |