Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KITLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000899.4 | NP_000890.1 | 4, 5 | 556 | ||
NM_003994.5 | NP_003985.2 | 4, 5 | 556 | |||
GenBank mRNA | AF119835.1 | - | 4, 5 | 503 | ||
AF400436.1 | - | 4, 5 | 544 | |||
AF400437.1 | - | 4, 5 | 503 | |||
AK290319.1 | - | 4, 5 | 552 | |||
AK293002.1 | - | 4, 5 | 472 | |||
BC069733.1 | - | 4, 5 | 517 | 7262647 | ||
BC069783.1 | - | 4, 5 | 518 | 7262659 | ||
BC069797.1 | - | 4, 5 | 513 | 7262576 | ||
BC074725.2 | - | 4, 5 | 401 | 30915176 | ||
BC126166.1 | - | 4, 5 | 511 | 8991882 | ||
BC143899.1 | - | 4, 5 | 511 | 9052417 | ||
CR749222.1 | - | 3, 4 | 304 | |||
M59964.1 | - | 4, 5 | 544 |