Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhebl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026967.4 | NP_081243.1 | 7, 8 | 674 | ||
GenBank mRNA | AK007708.1 | - | 7, 8 | 524 | ||
AK135858.1 | - | 7, 8 | 629 | |||
AY327413.1 | - | 7, 8 | 722 | |||
AY974592.1 | - | 3, 4 | 156 | |||
BC016521.1 | - | 7, 8 | 593 | 3487009 |