Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026983.2 | NP_081259.1 | 3 | 309 | ||
GenBank mRNA | AK007684.1 | - | 3 | 310 | ||
AK007800.1 | - | 3 | 230 | |||
AK131830.1 | - | 3 | 310 | |||
BC037376.1 | - | 2 | 145 | |||
BC104406.1 | - | 3 | 297 | 40050532 | ||
BC104407.2 | - | 3 | 297 | 40050533 |