Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081070.1 | NP_001074539.1 | 7 | 1075 | ||
GenBank mRNA | AK007743.1 | - | 7 | 1024 | ||
BC116671.1 | - | 7 | 1041 | 40107256 |