Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260511.1 | NP_001247440.1 | 2 | 194 | ||
NM_001260512.1 | NP_001247441.1 | 2 | 201 | |||
NM_001267598.1 | NP_001254527.1 | 2 | 201 | |||
NM_020300.4 | NP_064696.1 | 2 | 201 | |||
NM_145764.2 | NP_665707.1 | 2 | 184 | |||
NM_145791.2 | NP_665734.1 | 2 | 254 | |||
NM_145792.2 | NP_665735.1 | 2 | 176 | |||
NR_048545.1 | 2 | 201 | ||||
NR_048547.1 | 2 | 254 | ||||
XM_006719079.1 | XP_006719142.1 | 2 | 192 | |||
XM_011520673.1 | XP_011518975.1 | 2 | 192 | |||
XM_011520674.2 | XP_011518976.1 | 2 | 192 | |||
GenBank mRNA | AA314967.1 | - | 2 | 254 | ||
AK291148.1 | - | 2 | 190 | |||
AV655242.1 | - | 2 | 235 | |||
BC005923.1 | - | 2 | 169 | 4297300 | ||
BE568125.1 | - | 2 | 165 | |||
BG569388.1 | - | 2 | 185 | |||
BG617746.1 | - | 2 | 202 | |||
BG778603.1 | - | 2 | 202 | |||
BT006982.1 | - | 1 | 111 | |||
CR407629.1 | - | 1 | 111 | |||
H49331.1 | - | 2 | 199 | |||
J03746.1 | - | 2 | 184 | |||
R86750.1 | - | 2 | 176 |