Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260511.1 | NP_001247440.1 | 3 | 295 | ||
NM_001267598.1 | NP_001254527.1 | 3 | 302 | |||
NM_020300.4 | NP_064696.1 | 3 | 302 | |||
NM_145764.2 | NP_665707.1 | 3 | 285 | |||
NM_145791.2 | NP_665734.1 | 3 | 355 | |||
NM_145792.2 | NP_665735.1 | 3 | 277 | |||
GenBank mRNA | AK291148.1 | - | 3 | 291 | ||
AV655242.1 | - | 3 | 336 | |||
BC005923.1 | - | 3 | 270 | 4297300 | ||
BG569388.1 | - | 3 | 286 | |||
BG617746.1 | - | 3 | 303 | |||
BT006982.1 | - | 2 | 212 | |||
H49331.1 | - | 3 | 300 | |||
J03746.1 | - | 3 | 285 |