Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026993.3 | NP_081269.1 | 1, 2 | 474 | ||
GenBank mRNA | AK010430.1 | - | 1, 2 | 440 | ||
AK143507.1 | - | 1, 2 | 491 | |||
AK148907.1 | - | 1, 2 | 475 | |||
AK149185.1 | - | 1, 2 | 126 | |||
BC034505.1 | - | 1, 2 | 425 | 5134831 |