Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204366.1 | NP_001191295.1 | 3 | 434 | ||
NM_001204367.1 | NP_001191296.1 | 2 | 334 | |||
NM_002413.4 | NP_002404.1 | 3 | 434 | |||
XM_006714221.2 | XP_006714284.1 | 2 | 121 | |||
XM_011531972.2 | XP_011530274.1 | 2 | 100 | |||
XM_017008211.1 | XP_016863700.1 | 3 | 446 | |||
XM_017008212.1 | XP_016863701.1 | 3 | 446 | |||
XM_017008213.1 | XP_016863702.1 | 3 | 446 | |||
XM_017008214.1 | XP_016863703.1 | 2 | 121 | |||
GenBank mRNA | AA122237.1 | - | 3 | 434 | ||
AK130948.1 | - | 3 | 383 | |||
AK307231.1 | - | 2 | 169 | |||
BC025416.1 | - | 3 | 348 | 3937750 | ||
CR407640.1 | - | 3 | 182 | |||
DB257575.1 | - | 3 | 198 | |||
U77604.1 | - | 3 | 193 |