Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp746 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163475.1 | NP_001156947.1 | 6 | 981 | ||
GenBank mRNA | AK156312.1 | - | 6 | 982 |