Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004528.3 | NP_004519.1 | 3, 4 | 621 | ||
XM_005245174.3 | XP_005245231.2 | 3, 4 | 224 | |||
GenBank mRNA | AF026977.1 | - | 2, 3 | 242 | ||
AK223012.1 | - | 2, 3 | 221 | |||
AK312103.1 | - | 3, 4 | 237 | |||
BC000505.2 | - | 3, 4 | 220 | 2822075 | ||
BC003034.2 | - | 2, 3 | 202 | 2822075 | ||
BC005964.1 | - | 3, 4 | 270 | 4068898 | ||
BI666647.1 | - | 3, 4 | 286 | |||
CR450359.1 | - | 2, 3 | 193 |