Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027008.2 | NP_081284.2 | 3 | 482 | ||
GenBank mRNA | AK011637.1 | - | 3 | 217 | ||
AK052960.1 | - | 3 | 484 | |||
AK129077.1 | - | 3 | 442 | |||
AK143644.1 | - | 3 | 450 | |||
BC021777.1 | - | 3 | 434 | 4215964 |