Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023503.3 | NP_075992.2 | 2 | 890 | ||
GenBank mRNA | AB433625.1 | - | 2 | 393 | ||
AF078834.1 | - | 2 | 418 | |||
AK012716.1 | - | 2 | 887 | |||
AK048800.1 | - | 2 | 886 | |||
AK083144.1 | - | 2 | 889 | |||
BC050003.1 | - | 2 | 835 | |||
BC096433.1 | - | 2 | 870 | 6310342 | ||
BC138229.1 | - | 2 | 621 | 8861248 |