Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027016.2 | NP_081292.1 | 8 | 1362 | ||
GenBank mRNA | AK088093.1 | - | 8 | 911 | ||
AK090078.1 | - | 8 | 913 | |||
BC023720.1 | - | 5 | 577 | |||
BC053504.1 | - | 8 | 900 | |||
BC067202.1 | - | 8 | 897 | 6332701 | ||
BC071243.1 | - | 8 | 913 |