Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027016.2 | NP_081292.1 | 6 | 1067 | ||
GenBank mRNA | AK088093.1 | - | 6 | 616 | ||
AK090078.1 | - | 6 | 617 | |||
BC023720.1 | - | 3 | 282 | |||
BC053504.1 | - | 6 | 605 | |||
BC067202.1 | - | 6 | 602 | 6332701 | ||
BC071243.1 | - | 6 | 618 |