Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 3300002I08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027017.1 | NP_081293.4 | 4 | 450 | ||
GenBank mRNA | AK014369.1 | - | 4 | 450 | ||
BC132262.1 | - | 4 | 391 | 40130539 | ||
BC132264.1 | - | 4 | 391 | 40130541 |