Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhobtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081347.1 | NP_001074816.1 | 7 | 1650 | ||
NM_001252636.1 | NP_001239565.1 | 7 | 1633 | |||
NM_001252637.1 | NP_001239566.1 | 7 | 1754 | |||
NM_001252638.1 | NP_001239567.1 | 8 | 1813 | |||
GenBank mRNA | AK005770.1 | - | 7 | 1633 | ||
AK034826.1 | - | 7 | 1650 | |||
AK078306.1 | - | 6 | 1587 | |||
AK173015.1 | - | 2 | 1014 | |||
BC099973.1 | - | 7 | 1631 | 6432524 | ||
BC137646.1 | - | 6 | 1584 | 8860665 | ||
BC137647.1 | - | 6 | 1584 | 8860666 | ||
BC144800.1 | - | 7 | 1643 | 9053348 |