Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhobtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081347.1 | NP_001074816.1 | 5 | 486 | ||
NM_001252636.1 | NP_001239565.1 | 5 | 469 | |||
NM_001252637.1 | NP_001239566.1 | 5 | 590 | |||
NM_001252638.1 | NP_001239567.1 | 6 | 649 | |||
GenBank mRNA | AK005770.1 | - | 5 | 469 | ||
AK034826.1 | - | 5 | 486 | |||
AK078306.1 | - | 4 | 423 | |||
BC099973.1 | - | 5 | 467 | 6432524 | ||
BC137646.1 | - | 4 | 420 | 8860665 | ||
BC137647.1 | - | 4 | 420 | 8860666 | ||
BC144800.1 | - | 5 | 479 | 9053348 |