Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027030.2 | NP_081306.1 | 3, 4 | 543 | ||
GenBank mRNA | AK005584.1 | - | 3, 4 | 544 | ||
AK078253.1 | - | 3, 4 | 572 | |||
AK169641.1 | - | 3, 4 | 551 | |||
AK171018.1 | - | 3, 4 | 535 | |||
AY040775.1 | - | 3, 4 | 556 | |||
BC016273.1 | - | 3, 4 | 563 | 4511427 |