Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027030.2 | NP_081306.1 | 4 | 577 | ||
GenBank mRNA | AK005584.1 | - | 4 | 578 | ||
AK078253.1 | - | 4 | 606 | |||
AK169641.1 | - | 4 | 585 | |||
AK171018.1 | - | 4 | 569 | |||
AY040775.1 | - | 4 | 590 | |||
BC016273.1 | - | 4 | 597 | 4511427 |