Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172371.3 | NP_758959.1 | 4 | 1428 | ||
GenBank mRNA | AK005699.1 | - | 3 | 854 | ||
AK028143.1 | - | 4 | 1413 | |||
AK028759.1 | - | 4 | 1429 | |||
BC039949.1 | - | 4 | 1216 | 4167719 |