Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h4h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007642.1 | - | 1 | 431 | ||
BC028550.1 | - | 1 | 376 | 1398106 | ||
BC058529.1 | - | 1 | 372 | 6817839 | ||
BC087952.1 | - | 1 | 424 | 6787571 |