Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700025G04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197990.3 | NP_932107.1 | 2, 3 | 619 | ||
GenBank mRNA | AI552192.1 | - | 2, 3 | 268 | ||
AK006337.1 | - | 2, 3 | 277 | |||
AK148738.1 | - | 1, 2 | 136 | |||
AK167392.1 | - | 270 | ||||
BC034723.1 | - | 2, 3 | 398 | 3981517 | ||
BC055912.1 | - | 2, 3 | 434 | 3978655 |