Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002415.1 | NP_002406.1 | 2, 3 | 380 | ||
GenBank mRNA | AF469046.1 | - | 2, 3 | 283 | ||
AK311929.1 | - | 2, 3 | 375 | |||
BC000447.2 | - | 2, 3 | 364 | 2821346 | ||
BC007676.1 | - | 2, 3 | 348 | 3605756 | ||
BC008914.2 | - | 2, 3 | 334 | 3615896 | ||
BC013976.2 | - | 2, 3 | 379 | 4130651 | ||
BC022414.1 | - | 2, 3 | 381 | 4243461 | ||
BC053376.1 | - | 2, 3 | 317 | 6047427 | ||
BT007148.1 | - | 2, 3 | 283 | |||
CR407644.1 | - | 2, 3 | 283 | |||
CR456520.1 | - | 2, 3 | 303 | |||
CR541651.1 | - | 2, 3 | 283 | |||
JQ846015.1 | - | 2, 3 | 283 | |||
L10612.1 | - | 2, 3 | 357 | |||
M25639.1 | - | 2, 3 | 333 | |||
M95775.1 | - | 2, 3 | 257 | |||
Z23063.1 | - | 2, 3 | 283 |