Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700019M22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006132.1 | - | 1 | 99 | ||
BC050791.1 | - | 1 | 166 | 6742013 | ||
DQ309695.1 | - | 1 | 64 |