Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002416.2 | NP_002407.1 | 2 | 220 | ||
GenBank mRNA | AK311852.1 | - | 2 | 206 | ||
BC025745.1 | - | 2 | 195 | |||
BC042178.1 | - | 2 | 168 | |||
BC063122.1 | - | 2 | 211 | 30370826 | ||
BC095396.1 | - | 2 | 206 | 30336410 | ||
DB141449.1 | - | 2 | 222 | |||
X72755.1 | - | 2 | 185 |