Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufaf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027093.4 | NP_081369.2 | 7, 8 | 781 | ||
GenBank mRNA | AK009123.1 | - | 7, 8 | 848 | ||
BC005630.1 | - | 7, 8 | 648 |