Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027097.1 | NP_081373.1 | 4 | 640 | ||
GenBank mRNA | AK009217.1 | - | 3 | 503 | ||
BC147442.1 | - | 4 | 640 | 9056084 | ||
BC147463.1 | - | 4 | 640 | 9056105 |