Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172154.1 | NP_001165625.1 | 4 | 506 | ||
NM_027109.2 | NP_081385.1 | 4 | 623 | |||
GenBank mRNA | AK009174.1 | - | 4 | 629 | ||
AK150242.1 | - | 4 | 536 | |||
BC023246.1 | - | 4 | 506 | 4039470 | ||
DQ116784.1 | - | 4 | 517 |