Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045483.1 | NP_001038948.1 | 2 | 1393 | ||
NM_027115.2 | NP_081391.1 | 2 | 1289 | |||
GenBank mRNA | AK009250.1 | - | 2 | 1280 | ||
BC048375.1 | - | 2 | 1386 | |||
BC055699.1 | - | 3 | 1757 | |||
BC069850.1 | - | 1 | 957 | 5366053 |