Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027120.2 | NP_081396.1 | 4 | 360 | ||
GenBank mRNA | AK009352.1 | - | 4 | 376 | ||
AK137483.1 | - | 4 | 362 | |||
BC119244.1 | - | 4 | 322 | 8733993 | ||
BC119246.1 | - | 4 | 322 | 8733995 |