Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vkorc1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001327.2 | NP_001001327.1 | 2 | 342 | ||
NM_001286382.1 | NP_001273311.1 | 3 | 314 | |||
NM_027121.4 | NP_081397.1 | 3 | 452 | |||
GenBank mRNA | AK009497.1 | - | 2 | 222 | ||
AK078942.1 | - | 2 | 285 | |||
AY423046.1 | - | 3 | 394 | |||
BB625262.1 | - | 2 | 285 | |||
BC085133.1 | - | 2 | 342 | 6733526 | ||
BC125315.1 | - | 3 | 438 | 40129830 | ||
BC138132.1 | - | 3 | 438 | 8861151 | ||
CN701203.1 | - | 3 | 292 |