Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vkorc1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001327.2 | NP_001001327.1 | 2 | 837 | ||
NM_001286382.1 | NP_001273311.1 | 3 | 809 | |||
NM_027121.4 | NP_081397.1 | 3 | 947 | |||
GenBank mRNA | AK009497.1 | - | 2 | 717 | ||
AK078942.1 | - | 2 | 780 | |||
AY423046.1 | - | 3 | 889 | |||
BC085133.1 | - | 2 | 837 | 6733526 |