Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027122.3 | NP_081398.2 | 3, 4 | 1093 | ||
GenBank mRNA | AK009269.1 | - | 3, 4 | 1172 | ||
AK089539.1 | - | 3, 4 | 1094 | |||
BC019171.1 | - | 3, 4 | 1120 | 5003354 | ||
BC085305.1 | - | 3, 4 | 1157 | 6399056 |