Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKI67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145966.1 | NP_001139438.1 | 6 | 567 | ||
NM_002417.4 | NP_002408.3 | 6 | 567 | |||
XM_011539818.2 | XP_011538120.1 | 3 | 298 | |||
GenBank mRNA | AJ567756.1 | - | 3 | 177 | ||
X65550.1 | - | 6 | 568 | |||
X65551.1 | - | 6 | 568 |