Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145354.1 | NP_001138826.1 | 11 | 1232 | ||
NM_001321316.1 | NP_001308245.1 | 10 | 1163 | |||
NM_013255.4 | NP_037387.2 | 10 | 1160 | |||
XM_006715993.2 | XP_006716056.1 | 10 | 1160 | |||
XM_011516224.2 | XP_011514526.1 | 10 | 1160 | |||
GenBank mRNA | AF047489.1 | - | 10 | 1114 | ||
AK002024.1 | - | 10 | 1102 | |||
AK294384.1 | - | 11 | 1233 | |||
AK310452.1 | - | 10 | 1113 | |||
AL162071.1 | - | 1 | 140 | |||
BC002834.2 | - | 5 | 538 | |||
BC067825.1 | - | 10 | 1117 | 5268939 |