Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145131.1 | NP_660113.1 | 21, 22 | 2466 | ||
GenBank mRNA | AF513714.1 | - | 21, 22 | 2466 | ||
AK129292.1 | - | 21, 22 | 2440 | |||
AK141277.1 | - | 21, 22 | 2461 | |||
AK147837.1 | - | 21, 22 | 2457 | |||
AK164235.1 | - | 21, 22 | 2428 | |||
AK167426.1 | - | 21, 22 | 2457 | |||
AK168335.1 | - | 20, 21 | 2343 | |||
AY779273.1 | - | 21, 22 | 2466 | |||
AY779274.1 | - | 21, 22 | 2466 | |||
BC006917.1 | - | 11, 12 | 1267 | 3601081 |