Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027148.3 | NP_081424.3 | 5, 6 | 575 | ||
GenBank mRNA | AJ505005.1 | - | 5, 6 | 235 | ||
AK009584.1 | - | 5, 6 | 247 | |||
AK169719.1 | - | 3, 4 | 178 | |||
AK220227.1 | - | 5, 6 | 575 | |||
BC059089.1 | - | 5, 6 | 604 | 6509820 | ||
BC067250.1 | - | 4, 5 | 218 |