Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pir | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301402.1 | NP_001288331.1 | 5 | 645 | ||
NM_027153.3 | NP_081429.1 | 6 | 774 | |||
GenBank mRNA | AK009757.1 | - | 6 | 717 | ||
AK082922.1 | - | 6 | 659 | |||
AK134541.1 | - | 6 | 773 | |||
AK138314.1 | - | 6 | 770 | |||
AK151301.1 | - | 6 | 677 | |||
BC024062.1 | - | 6 | 680 | 5067536 |