Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197991.2 | NP_932108.2 | 4 | 503 | ||
GenBank mRNA | AK009801.1 | - | 4 | 413 | ||
AK150499.1 | - | 4 | 408 | |||
AK160274.1 | - | 4 | 418 | |||
AK169010.1 | - | 4 | 454 | |||
AK171657.1 | - | 4 | 417 | |||
AY761096.1 | - | 4 | 396 | |||
AY761098.1 | - | 4 | 396 | |||
BC006065.1 | - | 2 | 140 | |||
BC020179.1 | - | 4 | 398 | 4913500 |