Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178378.3 | NP_848465.1 | 11 | 1316 | ||
GenBank mRNA | AK010269.1 | - | 8 | 865 | ||
AK146422.1 | - | 9 | 1054 | |||
BC049990.1 | - | 11 | 1300 | 6310844 |