Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnaseh2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027187.3 | NP_081463.1 | 5 | 512 | ||
GenBank mRNA | AK010292.1 | - | 5 | 502 | ||
AK146518.1 | - | 5 | 513 | |||
AK220001.1 | - | 11 | ||||
BC029252.1 | - | 5 | 542 | 3666709 | ||
BC038158.1 | - | 5 | 586 | 1493985 |